Learn More
Sino Biological Recombinant Mouse SHP-1 Protein (aa 207-597, His & GST Tag)
Supplier:  Sino Biological 50894M20B100
Encompass_Preferred
A DNA sequence encoding the mouse PTPN6 (P29351-2) (Ala207-Lys597) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2604