Learn More
Sino Biological Recombinant Mouse BAMBI Protein (hFc Tag)
Supplier:  Sino Biological 50895M02H50
Encompass_Preferred
A DNA sequence encoding the mouse BAMBI (Q9D0L6) (Met1-Ala152) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2607