Learn More
Sino Biological Recombinant Mouse BAMBI Protein (His Tag)
Supplier:  Sino Biological 50895M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse BAMBI (Q9D0L6) (Met 1-Ala 152) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2609