Learn More
Perkin Elmer US LLC UV Quartz SUPRASIL Ultra-Micro Cell with Lid - 10 mm, 30 µL
Supplier:  Perkin Elmer US LLC B0631079
Encompass_Preferred
The UV Quartz SUPRASIL Ultra-Micro Cell with PTFE Lid has a light path of 10 mm, a chamber volume of 30 µL, two apertures of 1.5 mm, and outside dimensions of 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty . 1 NOTE: This cell is used only with the LAMBDA 365/465.
Catalog No. 50-904-2638