Learn More
Sino Biological Recombinant Mouse HMGB1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 50913M01H50
Encompass_Preferred
A DNA sequence encoding the mouse HMGB1 (P63158) (Met 1-Glu 215) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-2622