Learn More
Sino Biological Recombinant Mouse TROP-2 Protein (hFc Tag)
Supplier:  Sino Biological 50922M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TACSTD2 (Q8BGV3?) (Met 1-Gln 270) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2640