Learn More
Sino Biological Recombinant Mouse Contactin-1 Protein (His Tag)
Supplier:  Sino Biological 50933M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CNTN1 (NP_031753.1) (Met1-Leu1000) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2648