Learn More
Sino Biological Recombinant Mouse Nicastrin Protein (hFc Tag)
Supplier:  Sino Biological 50934M02H100
Encompass_Preferred
A DNA sequence encoding the mouse NCSTN (P57716) (Met1-Glu668) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2650