Learn More
Sino Biological Recombinant Mouse NAALADL1 Protein (His Tag)
Supplier:  Sino Biological 50937M07H50
Encompass_Preferred
A DNA sequence encoding the secreted form of mouse NAALADL1 (Q7M758) (Pro 29-Leu 745) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-2657