Learn More
Sino Biological Recombinant Mouse TIGIT Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50939M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TIGIT (NP_001139797.1) (Met 1-Gly 141) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2660