Learn More
Sino Biological Recombinant Mouse LRRTM4 Protein (ECD, His Tag)
Supplier:  Sino Biological 50944M08H50
Encompass_Preferred
A DNA sequence encoding the mouse Lrrtm4 (NP_848846.3) (Met1-Lys424) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2663