Learn More
Sino Biological Recombinant Mouse Vanin-1/VNN1 Protein (hFc Tag)
Supplier:  Sino Biological 50952M02H100
Encompass_Preferred
A DNA sequence encoding the mouse VNN1 (Q9Z0K8) (Met 1-Ser 487), without the pro peptide, was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2670