Learn More
Sino Biological Recombinant Mouse SPINK4 Protein (hFc Tag)
Supplier:  Sino Biological 50953M02H100
Encompass_Preferred
A DNA sequence encoding the mouse SPINK4 (NP_035593.2) (Met 1-Cys 86) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2674