Learn More
Sino Biological Recombinant Mouse MANF Protein (His Tag)
Supplier:  Sino Biological 50954M08H100
Encompass_Preferred
A DNA sequence encoding the mouse MANF (NP_083379.2) (Met 1-Leu 179) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2676