Learn More
Sino Biological Recombinant Mouse SIRP alpha/SHPS-1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50956M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse SIRPA (BAA20376.1) (Met1-Asn373) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2680