Learn More
Sino Biological Recombinant Mouse CNDP1 Protein (His Tag)
Supplier:  Sino Biological 50959M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CNDP1 (Q8BUG2) (Met 1-Tyr 492) was expressed, with an N-terminal signal peptide and a C-terminal polyhistidine tag.
Catalog No. 50-162-2686