Learn More
Sino Biological Recombinant Mouse GLA/alpha-Galactosidase A Protein (His Tag)
Supplier:  Sino Biological 50964M08H50
Encompass_Preferred
A DNA sequence encoding the mouse Gla (Q8BGZ6) (Met1-Arg421) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2695