Learn More
Sino Biological Recombinant Mouse NGFR/p75NTR Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 50971M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse NGFR (Q9Z0W1) (Met 1-Asn 253) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2700