Learn More
Sino Biological Recombinant Mouse B7-H3 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50973M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD276 (NP_598744.1) (Met1-Phe244) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2706