Learn More
Sino Biological Recombinant Mouse B7-H3 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50973M08HB100
Encompass_Preferred
A DNA sequence encoding the mouse CD276 (NP_598744.1) (Met1-Phe244) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-162-2708