Learn More
Sino Biological C1s-A subcomponent Protein, Mouse, Recombinant (His Tag)
Supplier:  Sino Biological 50979M08H100
Encompass_Preferred
A DNA sequence encoding the mouse C1sa (AAH18319.1) (Met1-Asp688) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2714