Learn More
Sino Biological Recombinant Mouse WIF1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50984M08H100
Encompass_Preferred
A DNA sequence encoding the mouse WIF1 (Q9WUA1) (Met1-Trp379) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2718