Learn More
Sino Biological Recombinant Mouse Cadherin-6/K-cadherin Protein (His Tag)
Supplier:  Sino Biological 50985M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CDH6 (P97326) (Met1-Ala615) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2720