Learn More
Sino Biological Recombinant Mouse Carboxypeptidase M Protein (His Tag)
Supplier:  Sino Biological 50990M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CPM isoform 1 (Q80V42-1) (Met 1-His 422), without the propeptide, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-162-2726