Learn More
Sino Biological Recombinant Mouse VEGFR2/KDR Protein (hFc Tag)
Supplier:  Sino Biological 50998M02H100
Encompass_Preferred
A DNA sequence encoding the mouse KDR (P35918) (Met1-Glu762) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2736