Learn More
Sino Biological Recombinant Mouse VEGFR2/KDR Protein (His Tag)
Supplier:  Sino Biological 50998M08H100
Encompass_Preferred
A DNA sequence encoding the mouse KDR (P35918-1) extracellular domain (Met 1-Glu 762) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2738