Learn More
Sino Biological Recombinant Mouse VEGFR2/KDR Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50998M08HB20
Encompass_Preferred
A DNA sequence encoding the mouse KDR (P35918-1) extracellular domain (Met1-Glu762) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-162-2740