Learn More
Andwin Scientific Container, Empty IV Bag, INTRAVIA™, PVC Ports, Non-DEHP, 50mL, 48/Case
SDP
Catalog No. NC9445237
Encompass_Preferred
The empty IntraVia plastic containers have PVC ports and have a volume capacity of 50 mL. The containers have the length of 9.75", width of 9.75", and height of 6". This product is sterile.
Catalog No. NC9445237 Supplier Andwin Scientific Supplier No. 2B8019
The empty IntraVia plastic containers have PVC ports and have a volume capacity of 50 mL. The containers have the length of 9.75", width of 9.75", and height of 6". This product is sterile.