Learn More
Trajan Scientific 50F-RTC/RSH-GT-8.5/0.63C 50UL
SDP
Catalog No. 502530337
Encompass_Preferred
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.02 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 3.3 in., 85 mm, Model: 50F-RTC/RSH-GT-8.5/0.63C, Volume: 50 uL, Connection Type: Fixed
Catalog No. 50-253-0337 Supplier Trajan Scientific Supplier No. 004864
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.02 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 3.3 in., 85 mm, Model: 50F-RTC/RSH-GT-8.5/0.63C, Volume: 50 uL, Connection Type: Fixed