Learn More
Perkin Elmer US LLC Pre-Cleaned Copper Tubing - 50 ft x 0.125 in O.D. x 0.065 in I.D.
Catalog No. 509045632
Encompass_Preferred
Full NIST traceability using established EPA and ASTM protocols by using the appropriate combination of permeation devices. The permeation tubes are certified to be permeating at a certain weigh and threshold (~+/– 2%). Uncertified tubes have an estimated rate and a very large threshold, up to + /– 25%.
Catalog No. 50-904-5632 Supplier Perkin Elmer US LLC Supplier No. N9300077
Full NIST traceability using established EPA and ASTM protocols by using the appropriate combination of permeation devices. The permeation tubes are certified to be permeating at a certain weigh and threshold (~+/– 2%). Uncertified tubes have an estimated rate and a very large threshold, up to + /– 25%.