Learn More
MSC Tower 30438303-01 3 Outlet, 125 Volt, 15 Amp Rating, 4 to 6mA Max Surge, Plug In, Automatic, Commercial, Industrial GFCI 50 Ft. Long Cord, 5-15P and (3)R NEMA Configuration, Polycarbonate Housing
Catalog No. NC1695465
Encompass
Tower 30438303-01 3 Outlet, 125 Volt, 15 Amp Rating, 4 to 6mA Max Surge, Plug In, Automatic, Commercial, Industrial GFCI 50 Ft. Long Cord, 5-15P and (3)R NEMA Configuration, Polycarbonate Housing
Catalog No. NC1695465 Supplier MSC Supplier No. 53159943
Tower 30438303-01 3 Outlet, 125 Volt, 15 Amp Rating, 4 to 6mA Max Surge, Plug In, Automatic, Commercial, Industrial GFCI 50 Ft. Long Cord, 5-15P and (3)R NEMA Configuration, Polycarbonate Housing