Learn More
The Fresh Test 50g Glucose Bottle - Plastic 30/pk, Non-GMO Dextrose
Catalog No. NC3627347 Shop All The Fresh Test Diagnostic Glucose Test Products
Encompass
The Fresh Test 50g Glucose Bottle - Plasticis a diagnostic glucose beverage used in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each bottle contains 50 grams of glucose in powder form. Patients should add water to the fill line, shake well, and consume within 5 minutes for a Glucose Load Test.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
BPA-free bottle
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Ordering Information
This catalog number represents 30 units per box. The Glucose Bottle - Plastic is Fresh Test's most popular product, trusted by laboratories and preferred by patients for its taste and ease of use.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.
Catalog No. NC3627347 Supplier The Fresh Test Supplier No. 271020500PB30BOX
The Fresh Test 50g Glucose Bottle - Plasticis a diagnostic glucose beverage used in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each bottle contains 50 grams of glucose in powder form. Patients should add water to the fill line, shake well, and consume within 5 minutes for a Glucose Load Test.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
BPA-free bottle
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Ordering Information
This catalog number represents 30 units per box. The Glucose Bottle - Plastic is Fresh Test's most popular product, trusted by laboratories and preferred by patients for its taste and ease of use.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.