Learn More
Ace Glass, Inc. Mantle, 50L, for Very fine spherical flask, 490mm OD x 228mmheight, (2)2000w-230v, includes terminal box, not rated
SDP
Catalog No. 50983198
Encompass
50L aluminum housed, table-model mantle for Schott spherical flask. 465mm I.D. x 228mm height. Max internal operating temperature is 450 Deg. centigrade. (3)1000w-115v, cULus rated. Includes welded aluminum terminal box for hard-wiring mantle.
Catalog No. 50-983-198 Supplier Ace Glass, Inc. Supplier No. 1205332
50L aluminum housed, table-model mantle for Schott spherical flask. 465mm I.D. x 228mm height. Max internal operating temperature is 450 Deg. centigrade. (3)1000w-115v, cULus rated. Includes welded aluminum terminal box for hard-wiring mantle.