Learn More
Ace Glass, Inc. Mantle, 50L, for 457mm diameter spherical flask, 457mm OD x 228mm height, (3)1000w-115v, includes terminal box d, .
SDP
Catalog No. 50977330
Encompass
50L aluminum housed, table-model mantle for 456mm diameter spherical flask. 457mm I.D. x 228mm height. Max internal operating temperature is 450 Deg. centigrade. Includes welded aluminum terminal box for hard-wiring mantle.
Catalog No. 50-977-330 Supplier Ace Glass, Inc. Supplier No. 1205331
50L aluminum housed, table-model mantle for 456mm diameter spherical flask. 457mm I.D. x 228mm height. Max internal operating temperature is 450 Deg. centigrade. Includes welded aluminum terminal box for hard-wiring mantle.