Learn More
Ace Glass, Inc. Aluminum housed Stirmantle for 50L Corning spherical flask. Independent variable speed stirring, ships complete with motor, stir control, stir bar & terminal box, 120v 50/60Hz
SDP
Catalog No. 50977310
Encompass
Aluminum housed Stirmantle for 50L Corning spherical flasks by Glas-Col. Features independent variable speed stirring. Ships complete with motor, stir control, stir bar, and terminal box. 120v 50/60Hz.
Catalog No. 50-977-310 Supplier Ace Glass, Inc. Supplier No. 1204550
Aluminum housed Stirmantle for 50L Corning spherical flasks by Glas-Col. Features independent variable speed stirring. Ships complete with motor, stir control, stir bar, and terminal box. 120v 50/60Hz.