Learn More
Chemglass Life Sciences 50mL Amber Centrifuge Tube, PS Rack, Sterile, 25/Pack
SDP
Catalog No. 501897804
Encompass_Preferred
Meet the requirements of analytical level. High strength non-toxic materials. Raw material complies with USP VI requirements. Printed clear graduations with marking area. PP for the tube and HDPE for the cap. One-hand operation design. Sterilized by E-Beam. DNase, RNase and pyrogen free. Maximum centrifugation speed 12,000 x g.
Catalog No. 50-189-7804 Supplier Chemglass Life Sciences Supplier No. CGN4303A50
Meet the requirements of analytical level. High strength non-toxic materials. Raw material complies with USP VI requirements. Printed clear graduations with marking area. PP for the tube and HDPE for the cap. One-hand operation design. Sterilized by E-Beam. DNase, RNase and pyrogen free. Maximum centrifugation speed 12,000 x g.