Learn More
Ace Glass, Inc. 50L CYL,JKTD FLASK,400MM FLNG
SDP
Catalog No. 5000854744
Encompass
Jacketed, cylindrical kilo scale reaction flasks featuring removable flush seal valves, beaded pipe jacket inlet/outlets and bottom drain. Double-sided graduations facilitate left or right hand orientation. 10 to 20L flasks have 200mm Duran style o-ring grooved flanges while larger flasks have 300 to 400mm KF style flat flanges. All flask sizes thru 50 liter have all glass lower seals, larger flasks incorporate silicone lower seals. Low aspect ratio saves hood space. Temp range: -60°C to 200°C. Max jacket pressure: 8 psig.
Catalog No. 50-008-54744 Supplier Ace Glass, Inc. Supplier No. 1285011
Jacketed, cylindrical kilo scale reaction flasks featuring removable flush seal valves, beaded pipe jacket inlet/outlets and bottom drain. Double-sided graduations facilitate left or right hand orientation. 10 to 20L flasks have 200mm Duran style o-ring grooved flanges while larger flasks have 300 to 400mm KF style flat flanges. All flask sizes thru 50 liter have all glass lower seals, larger flasks incorporate silicone lower seals. Low aspect ratio saves hood space. Temp range: -60°C to 200°C. Max jacket pressure: 8 psig.