Learn More
Chemglass Life Sciences 50L Process Reactor System, Squatty, Cylindrical, Jacketed, Glass, 400mm Flange, Brushless DC Electric Motor
SDP
Catalog No. 501531337
Encompass_Preferred
50L Process Reactor System, Squatty, Cylindrical, Jacketed, Glass, 400mm Flange, Brushless DC Electric Motor. 50L Squatty, Low Profile, glass reactor system with 400mm flange, on Chemglass's innovative Kwik Mount support frame. This reactor maximizes clearance under the drain valve. 400mm ID Flange Provides Wider Access For Insertion or Probes, Baffles and Other Accessories without Interfering with Agitators. Heavy Duty 2" Diameter Solid Stainless Steel Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability. Reactor Jacket Graduation Scale on Both Sides.
Catalog No. 50-153-1337 Supplier Chemglass Life Sciences Supplier No. CG1968W651
Add to Cart
Add to Cart
Request a Quote
50L Process Reactor System, Squatty, Cylindrical, Jacketed, Glass, 400mm Flange, Brushless DC Electric Motor. 50L Squatty, Low Profile, glass reactor system with 400mm flange, on Chemglass's innovative Kwik Mount support frame. This reactor maximizes clearance under the drain valve. 400mm ID Flange Provides Wider Access For Insertion or Probes, Baffles and Other Accessories without Interfering with Agitators. Heavy Duty 2" Diameter Solid Stainless Steel Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability. Reactor Jacket Graduation Scale on Both Sides.