Learn More
Foxx Life Sciences 50L PP Carboy, Non Sterile
GREENER_CHOICE
Catalog No. 5000846720
Encompass_Preferred
EZclear 50L Polypropylene PP Carboy is a translucent container that allows easy visual inspection of its contents. The single use 50L PP carboy maintains durability, chemical resistance, and autoclavability of standard polypropylene carboys. The 50L PP Wide mouth carboys features include large top handles, easy-to-read metric graduations (±5% certified accuracy). The 50L non sterile PP Carboys manufactured from USP Class VI, polypropylene resin, offering good chemical resistance and thermal stability. 50L Autoclavable PP carboys have a recommended sterilization cycle of 121 °C at 15 psi for 20 minutes.
Features:
VersaCap technology enables flexible, interchangeable connector options
Transparency with approximately 30% haze
Leak-proof cap design
Double bagged non sterile carboy
50L PP Round Carboys ideal for mixing
Made in Class 7 Cleanroom environment
Catalog No. 50-008-46720 Supplier Foxx Life Sciences Supplier No. 515EL51FLS
EZclear 50L Polypropylene PP Carboy is a translucent container that allows easy visual inspection of its contents. The single use 50L PP carboy maintains durability, chemical resistance, and autoclavability of standard polypropylene carboys. The 50L PP Wide mouth carboys features include large top handles, easy-to-read metric graduations (±5% certified accuracy). The 50L non sterile PP Carboys manufactured from USP Class VI, polypropylene resin, offering good chemical resistance and thermal stability. 50L Autoclavable PP carboys have a recommended sterilization cycle of 121 °C at 15 psi for 20 minutes.
Features:
VersaCap technology enables flexible, interchangeable connector options
Transparency with approximately 30% haze
Leak-proof cap design
Double bagged non sterile carboy
50L PP Round Carboys ideal for mixing
Made in Class 7 Cleanroom environment