Learn More
Chemglass Life Sciences Reactor Support Frame, Mobile, Jacketed, 50L, 400mm Flange
SDP
Catalog No. 501531343
Encompass_Preferred
Reactor Support Frame, Mobile, Jacketed, 50L, 400mm Flange. Open air design, with decreased frame depth, allows greater access to the reactor and peripheral glassware. Motor mount arm can be used with either electric, air, or XP motors and can swing out of the way for greater access to reactor. Design allows support frame to be used for 50L 400mm flange Jacketed reactor systems. Glassware can be quickly installed and removed by one person saving time and frustration normally associated with out-of-date designs.
Catalog No. 50-153-1343 Supplier Chemglass Life Sciences Supplier No. CG1968Y12
Add to Cart
Add to Cart
Request a Quote
Reactor Support Frame, Mobile, Jacketed, 50L, 400mm Flange. Open air design, with decreased frame depth, allows greater access to the reactor and peripheral glassware. Motor mount arm can be used with either electric, air, or XP motors and can swing out of the way for greater access to reactor. Design allows support frame to be used for 50L 400mm flange Jacketed reactor systems. Glassware can be quickly installed and removed by one person saving time and frustration normally associated with out-of-date designs.