Learn More
Chemglass Life Sciences Insulating Jacket for Jacketed Cylindrical 50L (Short / 300mm Flange) Reactor
SDP
Catalog No. 501539709
Encompass_Preferred
Similar to CG-10009 but Without the Heating Element. For use as a heat or cold shield on Chemglass jacketed reactors. Insulating jacket is made from a heavy fiberglass fabric on the inside with a silicone-coated fiberglass fabric on the outside. Mantle is slit lengthwise to a hole in the bottom center with lacing closure to accommodate a bottom drain valve.
Catalog No. 50-153-9709 Supplier Chemglass Life Sciences Supplier No. CG10011150
Similar to CG-10009 but Without the Heating Element. For use as a heat or cold shield on Chemglass jacketed reactors. Insulating jacket is made from a heavy fiberglass fabric on the inside with a silicone-coated fiberglass fabric on the outside. Mantle is slit lengthwise to a hole in the bottom center with lacing closure to accommodate a bottom drain valve.