Learn More
Hach Company Calibration Standard Solution 50 mg/L NH4-N for Amtax sc
Catalog No. NC1970376 Shop All Hach Company Products
Encompass
Calibration standard solution providing a certified 50 mg/L ammonium-nitrogen (NH4-N) concentration for use in calibrating online ammonium analyzers. Intended to support calibration across relevant measurement ranges and supplied with safety documentation accessible via the manufacturer's SDS portal.
  • Provides a certified 50 mg/L NH4-N concentration for calibration.
  • Ensures accurate and repeatable ammonium nitrogen measurements.
  • Ready to use to minimize preparation and handling time.
  • Includes safety documentation available through the manufacturer's SDS portal.
  • Supports traceable calibration across typical analyzer ranges.
Catalog No. NC1970376 Supplier Hach Company Supplier No. 2825854
Calibration standard solution providing a certified 50 mg/L ammonium-nitrogen (NH4-N) concentration for use in calibrating online ammonium analyzers. Intended to support calibration across relevant measurement ranges and supplied with safety documentation accessible via the manufacturer's SDS portal.
  • Provides a certified 50 mg/L NH4-N concentration for calibration.
  • Ensures accurate and repeatable ammonium nitrogen measurements.
  • Ready to use to minimize preparation and handling time.
  • Includes safety documentation available through the manufacturer's SDS portal.
  • Supports traceable calibration across typical analyzer ranges.