Learn More
Teknova 50mg/ml Chloramphenicol Solution, 25mL in 60mL Bottle, Sterile, 25Pk
SDP
Catalog No. 502746032
Encompass
50 mg/mL Chloramphenicol Solution in a bottle for laboratory use in microbial selection and cloning workflows. Chloramphenicol is a broad-spectrum inhibitor of prokaryotic protein synthesis that targets the 50S ribosomal subunit. This stock concentration is suitable for preparing working solutions to maintain chloramphenicol-selectable plasmids in bacterial cultures, including E. coli.
Common working concentrations are 25–34 µg/mL in agar or broth for E. coli and other chloramphenicol-selectable bacteria. Use aseptic technique when diluting into sterile media.
Catalog No. 50-274-6032 Supplier Teknova Supplier No. C038025PK
50 mg/mL Chloramphenicol Solution in a bottle for laboratory use in microbial selection and cloning workflows. Chloramphenicol is a broad-spectrum inhibitor of prokaryotic protein synthesis that targets the 50S ribosomal subunit. This stock concentration is suitable for preparing working solutions to maintain chloramphenicol-selectable plasmids in bacterial cultures, including E. coli.
Common working concentrations are 25–34 µg/mL in agar or broth for E. coli and other chloramphenicol-selectable bacteria. Use aseptic technique when diluting into sterile media.