Learn More
Teknova 50mg/mL Erythromycin Solution. 25mL in 60mL Bottle, Sterile.
SDP
Catalog No. 502388158
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-8158 Supplier Teknova Supplier No. E5325
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.