Learn More
Teknova 50mg/mL Kanamycin Solution. 1mL Tubes, 50 Tubes per Pack, Sterile.
SDP
Catalog No. NC0839150
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
This item is not returnable. View return policy
Catalog No. NC0839150 Supplier Teknova Supplier No. K8000
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.