Learn More
Worthington Biochemical Corporation Collagenase, Type 5, Filtered | ≥450 units per mg dry weight
SDP
Catalog No. NC1773629
Encompass
Collagenase Type 5 (Code: CLS-5), which is filtered through a 0.22 micron membrane and lyophilized in vials. Store at 2-8°C.
Catalog No. NC1773629 Supplier Worthington Biochemical Corporation Supplier No. LS005286
Collagenase Type 5 (Code: CLS-5), which is filtered through a 0.22 micron membrane and lyophilized in vials. Store at 2-8°C.