Learn More
Chemglass Life Sciences 50mL Tube, Reaction, 14/20 Inner Joint, 0-4mm Valve, Airfree, Schlenk
SDP
Catalog No. AF0538A02
Encompass_Preferred
50mL Tube, Reaction, 14/20 Inner Joint, 0-4mm Valve, Airfree, Schlenk. Cylindrical shaped tube useful for storage or simple reactions with a standard taper inner joint at the top and a Chem-Vac™ Chem-Cap valve sidearm. Plug may be removed for sampling from tube while a purge of inert gas is maintained through the sidearm. Valve size is as listed.
Catalog No. AF0538A02 Supplier Chemglass Life Sciences Supplier No. AF0538A02
50mL Tube, Reaction, 14/20 Inner Joint, 0-4mm Valve, Airfree, Schlenk. Cylindrical shaped tube useful for storage or simple reactions with a standard taper inner joint at the top and a Chem-Vac™ Chem-Cap valve sidearm. Plug may be removed for sampling from tube while a purge of inert gas is maintained through the sidearm. Valve size is as listed.