Learn More
Chemglass Life Sciences 50mL 3-Neck European Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20°
SDP
Catalog No. CG157002
Encompass_Preferred
50mL 3-Neck European Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20¿. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. With standard taper outer joints on center and side necks.
Catalog No. CG157002 Supplier Chemglass Life Sciences Supplier No. CG157002
50mL 3-Neck European Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20¿. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. With standard taper outer joints on center and side necks.