Learn More
Chemglass Life Sciences 50mL 3-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20°
SDP
Catalog No. CG155904
Encompass_Preferred
50mL 3-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20¿. Pear shape, HEAVY WALL 3-neck flask with standard taper outer joints on the center and 20¿ angled side necks.
Catalog No. CG155904 Supplier Chemglass Life Sciences Supplier No. CG155904
50mL 3-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20¿. Pear shape, HEAVY WALL 3-neck flask with standard taper outer joints on the center and 20¿ angled side necks.