Learn More
Ace Glass, Inc. Mantle, 50mL, for 48mm diameter spherical flask, 49mm ID x 23.9mm height, 60w-115v, CSA rated
SDP
Catalog No. 50983195
Encompass
50mL aluminum housed, table-model mantle for 48mm diameter spherical flask. 49mm I.D. x 23.9mm height. Maximum internal operating temperature is 450 Deg. centigrade. 60w-115v, cULus rated. Power cord included.
Catalog No. 50-983-195 Supplier Ace Glass, Inc. Supplier No. 1205305
50mL aluminum housed, table-model mantle for 48mm diameter spherical flask. 49mm I.D. x 23.9mm height. Maximum internal operating temperature is 450 Deg. centigrade. 60w-115v, cULus rated. Power cord included.